missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22259F
This item is not returnable.
View return policy
Description
DAP1 Polyclonal antibody specifically detects DAP1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| DAP1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| DAP1, DAP-1, death-associated protein, death-associated protein 1, MGC99796 | |
| The immunogen this antibody was made to, maps to a region between residue 52 and 102 of human Death-Associated Protein using the numbering given in entry NP_004385.1 (GeneID 1611). | |
| 0.1 mL | |
| Apoptosis, Cellular Markers | |
| 1611 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction