missing translation for 'onlineSavingsMsg'
Learn More

Dapsone Hydroxylamine, TRC

Product Code. 18003040
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18003040 5 mg 5mg
18022830 10 mg 10mg
18033830 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18003040 Supplier Toronto Research Chemicals Supplier No. TRCD1932605MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32695-27-5
Molecular Formula C12 H12 N2 O3 S
Molecular Weight (g/mol) 264.3
Synonym Dapsone-hydroxylamine, Aniline, N-hydroxy-4,4'-sulfonyldi- (8CI), 4-[(4-Aminophenyl)sulfonyl]-N-hydroxybenzenamine, 4-(Hydroxyamino)-4'-aminodiphenyl sulfone, 4-Amino-4'-(hydroxyamino)diphenyl sulfone, Dapsone hydroxylamine, N-Hydroxy-4,4'-sulfonyldi(aniline), Benzenamine, 4-[(4-aminophenyl)sulfonyl]-N-hydroxy-
IUPAC Name N-[4-(4-aminophenyl)sulfonylphenyl]hydroxylamine
SMILES Nc1ccc(cc1)S(=O)(=O)c2ccc(NO)cc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H12N2O3S/c13-9-1-5-11(6-2-9)18(16,17)12-7-3-10(14-15)4-8-12/h1-8,14-15H,13H2
Formula Weight 264.0569
Chemical Name or Material Dapsone Hydroxylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.