Learn More
Motic™ Darkfield Attachment
Product Code. 11761258
Change view
Click to view available options
For Use With (Equipment):
B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11761258 B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11761258 Supplier Motic™ Supplier No. 12220275
  • ONLINE EXCLUSIVE PRICE
    49.30€
    44.42€ / Pack of 1
    Save 4.88€
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enhance visual contrast using darkfield methodology. Motic™ Darkfield Attachment is a simple, plug-in type attachment.
  • Enhance contrast between specimen and background.
  • Simple, plug-in type attachment.
  • For use with: B1-223ASC, B3-223PL, DMWB1-223ASC microscopes.
Specifications
Type Darkfield Plug-in Attachment
For Use With (Equipment) B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes