missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Dasatinib

Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Dasatinib is a highly potent pan-Src/Bcr-Abl inhibitor (Ki values are 16 and 30 pM, respectively. Inhibits Bcr-Abl, Src, Lck, Fyn, c-kit and Yes with IC50 values in the subnanomolar range. Inhibits proliferation of tumor cells in vitro and exhibits anticancer acivity in vivo in a mouse chronic myelogenous leukemia (CML) xenograft model. Inhibits replication of SARS-CoV and MERS-CoV in vitro (EC50 values are 2.1and 5.5 μM, respectively). Dasatinib also enhances CD3-targeted lentiviral vector-mediated gene transfer by 3 to 10-fold.
Specifications
Specifications
| Chemical Name or Material | N-(2-Chloro-6-methylphenyl)-2-[[6-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-5-thiazolecarboxamide |
| CAS | 302962-49-8 |
| Synonym | BMS-354825 |
| Quantity | 100 mg |
| Molecular Formula | C22H26ClN7O2S |
| Formula Weight | 488.01 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction