missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBF4 Antibody (24D8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26632-50ul
This item is not returnable.
View return policy
Description
DBF4 Monoclonal antibody specifically detects DBF4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry
Specifications
| DBF4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 24D8 | |
| Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| ASKchif, DBF4 homolog (S. cerevisiae), DBF4ACHIF, DBF4-type zinc finger-containing protein 1, ZDBF1DBF-type containing 1 | |
| A synthesized peptide derived from Human DBF4 [UniProt Q9UBU7] | |
| 50 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 10926 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction