Learn More
DBU, TRC
Product Code. 30004063
Change view
Click to view available options
Quantity:
10 g
50 g
150 g
Unit Size:
10g
150g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004063 10 g 10g
30008923 50 g 50g
30008093 150 g 150g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004063 Supplier Toronto Research Chemicals Supplier No. TRCD41709010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6674-22-2
Molecular Formula C9 H16 N2
Molecular Weight (g/mol) 152.24
Synonym 1,8-Diazabicyclo[5.4.0]undec-7-ene, Pyrimido[1,2-a]azepine, 2,3,4,6,7,8,9,10-octahydro-, 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, 1,8-Diaza-7-bicyclo[5.4.0]undecene, 1,8-Diazabicyclo[5.4.0]undec-7-ene, 1,8-Diazabicyclo[5.4.0]undeca-7-ene, 1,8-Diazabicyclo[5.4.0]undecene-7, 1,8-Diazabicycloundec-7-ene, 2H,3H,4H,6H,7H,8H,9H,10H-Pyrimido[1,2-a]azepine, Active D 10, Alcanpoudre DBU 70-3KG, Amicure DBUE, DBU, Dabco DBU, Diazabicyclo[5.4.0]undec-7-ene, Lupragen N 700, NSC 111184, NSC 230466, PC CAT DBU, Polycat DBU, San-apro, U-CAT SA 851, DBU (GR53261X)
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCN2CCCN=C2CC1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C9H16N2/c1-2-5-9-10-6-4-8-11(9)7-3-1/h1-8H2
Formula Weight 152.1313
Percent Purity >95%
Chemical Name or Material 1,8-Diaza-7-bicyclo[5.4.0]undecene
Grade HPLC