missing translation for 'onlineSavingsMsg'
Learn More

DC-SIGN/CD209 Antibody (902404), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18592629 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18592629 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18592629 Supplier Novus Biologicals Supplier No. FAB8345MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen DC-SIGN/CD209
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 902404
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Host Species Rat
Immunogen NS0 mouse myeloma cell line transfected with mouse DC-SIGN/CD209, Met1-Lys238, Accession # Q91ZX1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 30835
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.