missing translation for 'onlineSavingsMsg'
Learn More

DCC Antibody (OTI4A1), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

256.00€ - 550.00€

Specifications

Antigen DCC
Clone OTI4A1
Dilution Western Blot 1:200
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306343
View Documents
Novus Biologicals
NBP3-44888-100ul
100 μL
550.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306209
View Documents
Novus Biologicals
NBP3-44888-30ul
30 μL
256.00€
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DCC Monoclonal antibody specifically detects DCC in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

DCC
Western Blot 1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC
Full length human recombinant protein of human DCC (NP_005206) produced in HEK293T cell.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
OTI4A1
Western Blot
Unconjugated
Mouse
Apoptosis, Cancer, Tumor Suppressors
PBS (pH 7.3) 1% BSA, 50% glycerol
1630
IgG2b
>95%
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.