Learn More
DCDC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26978-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization. Mutations in this gene have been associated with Reading Disability, also referred to as developmental dyslexia. Alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
| DCDC2 | |
| Polyclonal | |
| Unconjugated | |
| DCDC2 | |
| DCDC2, DCDC2A, KIAA1154, Protein RU2S, RU2, RU2S | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 195208, 51473 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5DU00, Q9UHG0 | |
| DCDC2 | |
| DCDC2 Fusion Protein Ag25577 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |