Learn More
DCIR/CLEC4A Antibody (216110) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1748K
Product Code. 18542955
  • 712.00€ / 0.10mL
Description
DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DCIR/CLEC4A
Monoclonal
DyLight 488
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
216110
Western Blot, Flow Cytometry, CyTOF-ready
CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor
Mouse myeloma cell line NS0-derived recombinant human DCIR isoform 1, Gln70-Leu237, Accession # Q9UMR7
0.1 mL
Immunology, Innate Immunity
50856
Store at 4°C in the dark.