Learn More
DCP1B Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809930S
Encompass_Preferred
 View more versions of this product
Product Code. 17522604
  • 242.00€ / 30µL
Description
DCP1B Monoclonal antibody specifically detects DCP1B in Human samples. It is validated for Western Blot
Specifications
Specifications
DCP1B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8IZD4
Mouse
Affinity chromatography
RUO
196513
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1G4
Unconjugated
DCP1B
B930050E02Rik; DCP1; DCP1 decapping enzyme homolog b; DCP1 decapping enzyme homolog B (S. cerevisiae); Dcp1b; decapping enzyme hDcp1b; decapping mRNA 1B; hDcp1b; mRNA-decapping enzyme 1B; RGD1562214
Human recombinant protein fragment corresponding to amino acids 167-473 of human DCP1B produced in E.coli.