missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488 Anti-Mouse CD40L/CD154 (MR1), Proteintech
Hamster Monoclonal Antibody
Brand: Proteintech CL488-65129-100TESTS
This item is not returnable.
View return policy
Description
The CD40L/CD154 antibody from Proteintech is a armenian hamster monoclonal antibody generated with n/a of mouse CD40L/CD154. This antibody recognizes mouse antigen. The CD40L/CD154 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 488.Specifications
| CD40L/CD154 | |
| Monoclonal | |
| 500 μg/mL | |
| Armenian Hamster | |
| 100 Tests | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG |
| Flow Cytometry | |
| MR1 | |
| CoraLite Plus 488 | |
| Affinity purification | |
| RUO | |
| 21947 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction