Learn More
DDR2 Antibody (290804), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB25381MFV610
Product Code. 18542729
  • 718.00€ / 0.10mL
Description
DDR2 Monoclonal antibody specifically detects DDR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DDR2
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction
4921
Store at 4C in the dark.
Flow Cytometry, Immunohistochemistry, CyTOF
290804
Flow Cytometry, Immunohistochemistry, CyTOF-reported
CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase
Mouse myeloma cell line NS0-derived recombinant human DDR2, Gln24-Arg399, Accession # Q16832
RUO
Primary
Human
IgG2b