missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DDX11 293T Cell Transient Overexpression Lysate

Product Code. 18362388 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362388 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362388 Supplier Novus Biologicals™ Supplier No. H00001663T01
  • ONLINE EXCLUSIVE PRICE
    499.00€
    471.45€ / 0.01mL
    Save 27.55€
    5% Off
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Research Category Cell Biology, Cell Cycle and Replication
Target DDX11
Synonym CHL1CHL1-related helicase gene-1, CHL1-related protein 1, CHLR1MGC9335, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S.cerevisiae), DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (S.cerevisiae CHL1-likehelicase), DEAD/H box protein 11, EC 3.6.1, EC 3.6.1.23, EC 3.6.1.7, EC 3.6.4.13, hCHLR1, Keratinocyte growth factor-regulated gene 2 protein, KRG-2, KRG2CHL1-like helicase homolog, MGC133249, probable ATP-dependent RNA helicase DDX11
Gene Symbol DDX11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.