Learn More
DDX5 Mouse anti-Human, Unconjugated, Clone: 2D3D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67025-1-IG-20UL
Description
The DDX5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX5. This antibody recognizes human, mouse, rat antigen. The DDX5 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| DDX5 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2D3D2 | |
| Unconjugated | |
| 20 μL | |
| 1655 | |
| IgG1 |