missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX53 Antibody (CL7899) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88909CL1
This item is not returnable.
View return policy
Description
DDX53 Monoclonal antibody specifically detects DDX53 in Human samples. It is validated for Western Blot
Specifications
| DDX53 | |
| Monoclonal | |
| CoraFluor 1 | |
| CAGEprobable ATP-dependent RNA helicase DDX53, Cancer/testis antigen 26, Cancer-associated gene protein, CT26EC 3.6.4.13, DEAD (Asp-Glu-Ala-Asp) box polypeptide 53, DEAD box protein 53, DEAD box protein CAGE, DEAD-box protein CAGE, EC 3.6.1 | |
| This antibody was developed using a synthetic peptide derived from Q86TM3, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 168400 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| CL7899 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction