missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14632-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.Specifications
| DDX6 | |
| Polyclonal | |
| Unconjugated | |
| DDX6 | |
| ATP dependent RNA helicase p54, DDX6, DEAD box protein 6, HLR2, Oncogene RCK, P54, RCK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13209, 1656, 500988 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26196, P54823 | |
| Ddx6 | |
| DDX6 Fusion Protein Ag6211 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction