missing translation for 'onlineSavingsMsg'
Learn More

Deacetylcephalothin (>90%), TRC

Product Code. 30351193
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351193 25 mg 25mg
30354774 10 mg 10mg
30355046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351193 Supplier Toronto Research Chemicals Supplier No. TRCD19808525MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5935-65-9
Molecular Formula C14H14N2O5S2
Molecular Weight (g/mol) 354.4
Synonym (6R,7R)-3-(Hydroxymethyl)-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, (6R-trans)-3-(Hydroxymethyl)-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, 3-(Hydroxymethyl)-8-oxo-7-[2-(2-thienyl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, (6R,7R)-3-(Hydroxymethyl)-8-oxo-7-[[2-(2-thienyl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, (6R,7R)-3-Hydroxymethyl-7-(thien-2-ylacetamido)ceph-3-em-4-carboxylic Acid, 3-Hydroxymethyl-7-(2-thienylacetamide)-3-cephem-4-carboxylic Acid, Deacetylcephalotin, Desacetylcephalothin, Cefalotin EP Impurity B
IUPAC Name (6R,7R)-3-(hydroxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES OCC1=C(N2[C@H](SC1)[C@H](NC(=O)Cc3cccs3)C2=O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H14N2O5S2/c17-5-7-6-23-13-10(12(19)16(13)11(7)14(20)21)15-9(18)4-8-2-1-3-22-8/h1-3,10,13,17H,4-6H2,(H,15,18)(H,20,21)/t10-,13-/m1/s1
Formula Weight 354.0344
Chemical Name or Material Deacetylcephalothin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.