missing translation for 'onlineSavingsMsg'
Learn More

DEC2/SHARP1 Antibody (OTI5B1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-72203APC

Product Code. 30029798

  • 740.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DEC2/SHARP1 Monoclonal antibody specifically detects DEC2/SHARP1 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DEC2/SHARP1
Monoclonal
APC
PBS
Mouse
Immunogen affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry
OTI5B1
Western Blot, Immunohistochemistry
basic helix-loop-helix domain containing, class B, 3, basic helix-loop-helix family, member e41, bHLHb3, BHLHB3class E basic helix-loop-helix protein 41, bHLHe41, Class B basic helix-loop-helix protein 3, DEC2Enhancer-of-split and hairy-related protein 1, Differentially expressed in chondrocytes protein 2, SHARP-1, SHARP1hDEC2
Human recombinant protein fragment corresponding to amino acids 1-297 of human BHLHE41(NP_110389) produced in E.coli.
0.1 mL
Cancer, Endocrinology, Neuroscience, Signal Transduction
79365
Store at 4C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.