missing translation for 'onlineSavingsMsg'
Learn More

Decafluorobiphenyl, 99%

Product Code. 11478496
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478496 5 g 5g
11488496 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478496 Supplier Thermo Scientific Alfa Aesar Supplier No. A15948.06
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 434-90-2 | C12F10 | 334.116 g/mol

Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-90-2
Molecular Formula C12F10
Molecular Weight (g/mol) 334.116
MDL Number MFCD00000292
InChI Key ONUFSRWQCKNVSL-UHFFFAOYSA-N
Synonym decafluorobiphenyl, perfluorobiphenyl, biphenyl, decafluoro, perfluorodiphenyl, 1,1'-biphenyl, 2,2',3,3',4,4',5,5',6,6'-decafluoro, 1,2,3,4,5-pentafluoro-6-2,3,4,5,6-pentafluorophenyl benzene, decafluoro biphenyl, pubchem9114, acmc-1ctuw
PubChem CID 67949
IUPAC Name 1,2,3,4,5-pentafluoro-6-(2,3,4,5,6-pentafluorophenyl)benzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 66°C to 70°C
Density 1.785
Boiling Point 206°C
Quantity 5 g
Beilstein 1916978
Solubility Information Insoluble in water.
Formula Weight 334.12
Percent Purity 99%
Chemical Name or Material Decafluorobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.