missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decanoic-9,9,10,10,10-d5 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Decanoic-9,9,10,10,10-d5 Acid |
| CAS | 1219803-00-5 |
| Molecular Formula | C10 D5 H15 O2 |
| Quantity | 0.5 g |
| Synonym | 9,9,10,10,10-Pentadeuteriodecanoic acid, Decanoic acid-9,9,10,10,10-d5 |
| InChI Formula | InChI=1 S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCCCC(=O)O |
| IUPAC Name | 9,9,10,10,10-pentadeuteriodecanoic acid |
| Molecular Weight (g/mol) | 177.3 |
| Formula Weight | 177.1777 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction