Learn More
Fisherbrand™ Decapping Heads for Programmable Electronic High Power Crimp Station
Product Code. 15552310 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Diameter (Metric) Cap:
11 mm
20 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Cap Unit Size
15552310 11 mm Each
15542340 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15552310
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Programmable Electronic High Power Crimp Station
  • Decapping Head for Alumnium/Steel Caps
  • Not adjustable
TRUSTED_SUSTAINABILITY
Specifications
Description Decapping Head
Diameter (Metric) Cap 11 mm
Material Plastic, Steel
For Use With (Equipment) 11 mm Alumnium Caps
Autoclavable Not Autoclavable
Product Type Decapping Head