missing translation for 'onlineSavingsMsg'
Learn More

Dechloro-Rivaroxaban, TRC

Product Code. 30133199
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133199 1 g 1g
30008418 50 mg 50mg
30005218 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133199 Supplier Toronto Research Chemicals Supplier No. TRCD2266101G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1415566-28-7
Molecular Formula C19 H19 N3 O5 S
Molecular Weight (g/mol) 401.44
Synonym N-[[(5S)-2-Oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-2-thiophenecarboxamide, Dechloro-Rivaroxaban
IUPAC Name N-[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide
SMILES O=C(NC[C@H]1CN(C(=O)O1)c2ccc(cc2)N3CCOCC3=O)c4cccs4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H19N3O5S/c23-17-12-26-8-7-21(17)13-3-5-14(6-4-13)22-11-15(27-19(22)25)10-20-18(24)16-2-1-9-28-16/h1-6,9,15H,7-8,10-12H2,(H,20,24)/t15-/m0/s1
Formula Weight 401.1045
Percent Purity >95%
Chemical Name or Material N-[[(5S)-2-Oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-2-thiophenecarboxamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.