Learn More
β-Decitabine-3',5'-bis(4-chlorobenzoate), TRC
Product Code. 30326966
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326966 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326966 Supplier Toronto Research Chemicals Supplier No. TRCA601705250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Amino-1-[3,5-bis-O-(4-chlorobenzoyl)-2-deoxy-beta-D-erythro-pentofuranosyl]-1,3,5-triazin-2(1H)-one
CAS 1034301-08-0
Molecular Formula C22 H18 Cl2 N4 O6
Quantity 250 mg
Synonym 4-Amino-1-[3,5-bis-O-(4-chlorobenzoyl)-2-deoxy-b-D-erythro-pentofuranosyl]-1,3,5-triazin-2(1H)-one, β-Decitabine-3',5'-bis(4-chlorobenzoate), Decitabine USP Related Compound B
InChI Formula InChI=1S/C22H18Cl2N4O6/c23-14-5-1-12(2-6-14)19(29)32-10-17-16(34-20(30)13-3-7-15(24)8-4-13)9-18(33-17)28-11-26-21(25)27-22(28)31/h1-8,11,16-18H,9-10H2,(H2,25,27,31)/t16-,17+,18+/m0/s1
SMILES NC1=NC(=O)N(C=N1)[C@H]2C[C@H](OC(=O)c3ccc(Cl)cc3)[C@@H](COC(=O)c4ccc(Cl)cc4)O2
IUPAC Name [(2R,3S,5R)-5-(4-amino-2-oxo-1,3,5-triazin-1-yl)-3-(4-chlorobenzoyl)oxyoxolan-2-yl]methyl 4-chlorobenzoate
Molecular Weight (g/mol) 505.308
Formula Weight 504.06