missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NALP10 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142882
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP10 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. It was initially identified as PYNOD, an Apaf-1-like protein that binds to ASC, caspase-1 and IL-1-b, inhibiting the autoprocessing of caspase-1, caspase-1-mediated IL-1-b processing, and ASC aggregation. NALP10 is thus a potent regulator of apoptosis and inflammation.
Specifications
| NALP10 | |
| Polyclonal | |
| Unconjugated | |
| NLRP10 | |
| 6430548I20Rik; CLR11.1; NACHT, leucine rich repeat and PYD containing 10; NACHT, LRR and PYD containing protein 10; NACHT, LRR and PYD domains-containing protein 10; NACHT/LRR/pyrin domain-containing protein 10; NALP10; Napl10; neuronal apoptosis inhibitor protein 10; NLR family pyrin domain containing 10; NLR family, pyrin domain containing 10; NLRP10; NOD8; Nucleotide-binding oligomerization domain protein 8; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 10; PAN5; Pynod | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 244202 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8CCN1 | |
| NLRP10 | |
| Peptide with sequence NDLEENSFKTLKFH-C (aa 18-31). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction