Learn More
Borer Chemie™ deconex 22 LIQ-x
Product Code. 17445122
Change view
Click to view available options
Quantity:
1 x 6 kg
1 x 30 kg
Unit Size:
30kg
6kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17445122 1 x 6 kg 6kg
17444922 1 x 30 kg 30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17445122 Supplier Borer Chemie™ Supplier No. 519200.00KK5W
Universal use for automated cleaning of laboratory glassware and utensils. Without phosphates, surfactants and chlorine. deconex 22 LIQ-x is an ecological solution with an excellent performance. Acidic rinse with a deconex product recommended.
  • Concentrate for universal automated cleaning
  • Ecological solution without phosphates and surfactants
  • Without chlorine, EDTA or NTA
  • High yield, typical concentration: 5-10 mL/L
  • Broad cleaning spectrum
  • High performance thanks to high dispersing capacity
  • Can be rinsed off without leaving residues
  • For consistent and flawless results
  • Acid rinse with a deconex cleaner recommended
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼12.2 (1% Solution)
Composition Alkalis, complexing agents, sequestering agents
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 6 kg
Volume (English) 1.3 gal.
Volume (Metric) 4.9 L
Weight (English) 13.2 lb.
Weight (Metric) 6 kg
Show More Show Less
compliance-icons
Product Identifier
  • 6KG deconex 22 LIQ-x
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide,dipotassium trioxosilicate