missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA185833
This item is not returnable.
View return policy
Description
Sheep anti human decorin antibody recognizes the C-terminal region of human bone proteoglycan II, also known as decorin, PG-S2 or PG40.
Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.
Specifications
| Decorin | |
| Polyclonal | |
| Unconjugated | |
| DCN | |
| Bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; Dermatan sulphate proteoglycans II (DSPG2); DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; Small leucine rich protein 1B; small leucine-rich protein 1B; unnamed protein product | |
| Sheep | |
| Protein G | |
| RUO | |
| 1634, 280760, 417892 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P07585, P21793, P28675 | |
| DCN | |
| Synthetic peptide, KLH conjugated, corresponding to residues C(316) V Y V R S A I Q L G N Y K(329) of Bone Proteoglycan II Precursor. | |
| 1 mL | |
| Primary | |
| Bovine, Chicken, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction