missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin-1/CLEC7A Antibody (BD6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
198.00€ - 523.00€
Specifications
| Antigen | Dectin-1/CLEC7A |
|---|---|
| Clone | BD6 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:25 - 1:200 |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203653
|
Novus Biologicals
NBP1-21513-0.025MG |
0.025 mg |
198.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18217640
|
Novus Biologicals
NBP1-21513 |
0.1 mg |
523.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Human samples. It is validated for Flow Cytometry.Specifications
| Dectin-1/CLEC7A | |
| 1.0 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| Q9BXN2 | |
| 64581 | |
| NIH3T3 cells expressing full length Beta-glucan receptor | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| BD6 | |
| Flow Cytometry 1:25 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
| CLEC7A | |
| IgG2b | |
| Protein G purified | |
| NBP1-21513 recognizes human Dectin-1, also known as beta-glucan receptor (betaGR), a C-type lectin domain family member. It is a major pathogen pattern-recognition receptor that binds beta-glucans, glucose polymers that form cell-wall components or exopolymers of yeasts, fungi and some bacteria. Beta-glucans have also been used experimentally and therapeutically as immunomodulators that enhance resistance to bacterial, yeast, viral and protozoan infections, as well as tumor formation. Dectin-1 also acts as a co-stimulatory molecule on T-cells to induce their proliferation and is necessary for the TLR2-mediated inflammatory response. It is highly expressed on peripheral blood leukocytes and dendritic cells. Human Dectin-1 has two major (betaGR-A and betaGR-B) and several minor isoforms. betaGR-A and betaGR-B differs by the presence and absence, respectively, of a stalk region. Clone BD6 recognizes betaGR-A only. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title