missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin-2/CLEC6A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03472-0.1MG
This item is not returnable.
View return policy
Description
Dectin-2/CLEC6A Polyclonal specifically detects Dectin-2/CLEC6A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Dectin-2/CLEC6A | |
| Polyclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
| Rabbit | |
| IgG purified | |
| RUO | |
| 93978 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| LYOPH | |
| Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml | |
| Q6EIG7 | |
| CLEC6A | |
| A synthetic peptide from n-terminal region of human Dectin-2 (CLEC6A, Dectin 2, Dectin2) conjugated to an immunogenic carrier protein was used as the antigen. | |
| 0.1 mg | |
| Primary | |
| Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction