missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethylammonium Bromide, TRC

Product Code. 30370025
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370025 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370025 Supplier Toronto Research Chemicals Supplier No. TRCD525538250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Decyltrimethylammonium Bromide
CAS 2082-84-0
Molecular Formula C13 H30 N . Br
Quantity 250 mg
Synonym 1-Decanaminium, N,N,N-trimethyl-, bromide (1:1) (ACI), 1-Decanaminium, N,N,N-trimethyl-, bromide (9CI), Ammonium, decyltrimethyl-, bromide (8CI), Decyltrimethylammonium bromide (6CI, 7CI), DeTAB, DTAB, FSM 20, N,N,N-Trimethyl-1-decanaminium bromide, N,N,N
InChI Formula InChI=1S/C13H30N.BrH/c1-5-6-7-8-9-10-11-12-13-14(2,3)4;/h5-13H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCCCCCCCCC[N+](C)(C)C
IUPAC Name decyl(trimethyl)azanium;bromide
Molecular Weight (g/mol) 280.288
Formula Weight 279.156
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.