missing translation for 'onlineSavingsMsg'
Learn More

DEET omega-Carboxylic Acid, TRC

Product Code. 30137165
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137165 100 mg 100mg
30134220 250 mg 250mg
30008744 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137165 Supplier Toronto Research Chemicals Supplier No. TRCD228505100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72236-23-8
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.25
Synonym 3-[(Diethylamino)carbonyl]benzoic Acid
IUPAC Name 3-(diethylcarbamoyl)benzoic acid
SMILES OC(C1=CC(C(N(CC)CC)=O)=CC=C1)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H15NO3/c1-3-13(4-2)11(14)9-6-5-7-10(8-9)12(15)16/h5-8H,3-4H2,1-2H3,(H,15,16)
Formula Weight 221.1052
Chemical Name or Material DEET omega-Carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.