Learn More
OHAUS™ Defender™ 3000 Washdown - I-D33 Bench Scale (EU)
Product Code. 17585815 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
15 kg
30 kg
60 kg
150 kg
300 kg
Model:
D33XW150C1L7
D33XW150C1X7
D33XW15C1R6
D33XW300C1X7
D33XW30C1R6
D33XW60C1L7
D33XW60C1R6
Readability:
2 g
5 g
10 g
20 g
50 g
Dimensions (L x W x H):
491 x 305 x 569 mm
688 x 420 x 922 mm
787 x 500 x 922 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Model Readability Dimensions (L x W x H) Unit Size
17585815 300 kg D33XW300C1X7 50 g 787 x 500 x 922 mm Each
17525815 15 kg D33XW15C1R6 2 g 491 x 305 x 569 mm Each
17535815 30 kg D33XW30C1R6 5 g 491 x 305 x 569 mm Each
17545815 60 kg D33XW60C1R6 10 g 491 x 305 x 569 mm Each
17555815 60 kg D33XW60C1L7 10 g 688 x 420 x 922 mm Each
17565815 150 kg D33XW150C1L7 20 g 688 x 420 x 922 mm Each
17575815 150 kg D33XW150C1X7 20 g 787 x 500 x 922 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17585815 Supplier OHAUS™ Supplier No. 30684781
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Stainless Steel Bench Scales for Basic Industrial Washdown Applications
  • Results are clearly shown using the large, bright backlit LCD display.
  • In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
  • This industrial scale features a 304 stainless steel platform and frame, IP67 load cell.
  • Indicator features an IP66 washdown protected 304-stainless steel housing with mounting bracket.
  • Features software for Parts Counting, Checkweighing and Totalization.
  • Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 100 Hours with Rechargeable Battery
Battery Type AC Adapter (Included);Rechargeable Battery (Included)
Connection Type Standard RS232 and optional USB Device or Ethernet
Construction 304-grade stainless steel platform and frame, stainless steel IP66 indicator, IP67 load cell, non-slip adjustable rubber feet
Capacity 300 kg
Display Type 7-segment Multi-color Backlit LCD
Dimensions (LxW) Platform 650 x 500 mm
Readability 50 g
Model D33XW300C1X7
Protection Class IP66
Temperature (Metric) Operating -10°C to +40°C, 85%RH, non-condensing
Weighing Modes Weighing, Checkweighing, Counting, Totalization.
Dimensions (L x W x H) 787 x 500 x 922 mm
Weight (Metric) 31.8 kg