missing translation for 'onlineSavingsMsg'
Learn More

Defluoro Atorvastatin Calcium Salt, TRC

Product Code. 30008410
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008410 50 mg 50mg
30008139 100 mg 100mg
30005141 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008410 Supplier Toronto Research Chemicals Supplier No. TRCD28987550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433289-83-9
Molecular Formula 2 C33 H35 N2 O5 . Ca
Molecular Weight (g/mol) 1119.36
Synonym 1H-Pyrrole-1-heptanoic acid, β,δ-dihydroxy-2-(1-methylethyl)-4,5-diphenyl-3-[(phenylamino)carbonyl]-, calcium salt (2:1), (βR,δR)-, Desfluoroatorvastatin Calcium Salt
IUPAC Name calcium;(3R,5R)-7-[2,3-diphenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
SMILES [Ca+2].CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccccc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)[O-].CC(C)c5c(C(=O)Nc6ccccc6)c(c7ccccc7)c(c8ccccc8)n5CC[C@@H](O)C[C@@H](O)CC(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/2C33H36N2O5.Ca/c2*1-22(2)31-30(33(40)34-25-16-10-5-11-17-25)29(23-12-6-3-7-13-23)32(24-14-8-4-9-15-24)35(31)19-18-26(36)20-27(37)21-28(38)39;/h2*3-17,22,26-27,36-37H,18-21H2,1-2H3,(H,34,40)(H,38,39);/q;;+2/p-2/t2*26-,27-;/m11./s1
Formula Weight 1118.4718
Chemical Name or Material (3R,5R)-7-[3-(phenylcarbamoyl)-2-isopropyl-4,5-diphenyl-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoic acid calcium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.