missing translation for 'onlineSavingsMsg'
Learn More

Defluoro Linezolid, TRC

Product Code. 30136787
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136787 10 mg 10mg
30005301 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136787 Supplier Toronto Research Chemicals Supplier No. TRCD22880010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556801-15-1
Molecular Formula C16H21N3O4
Molecular Weight (g/mol) 319.36
Synonym Acetamide, N-[[(5S)-3-[4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]-, Desfluorolinezolid
IUPAC Name N-[[(5S)-3-(4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide
SMILES CC(=O)NC[C@H]1CN(C(=O)O1)c2ccc(cc2)N3CCOCC3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H21N3O4/c1-12(20)17-10-15-11-19(16(21)23-15)14-4-2-13(3-5-14)18-6-8-22-9-7-18/h2-5,15H,6-11H2,1H3,(H,17,20)/t15-/m0/s1
Formula Weight 319.153
Percent Purity >95%
Chemical Name or Material Defluoro Linezolid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.