missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Norketamine, TRC

Product Code. 30011506
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011506 1 mg 1mg
30008115 5 mg 5mg
30010023 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011506 Supplier Toronto Research Chemicals Supplier No. TRCD2299501MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57683-62-2
Molecular Formula C12 H12 Cl N O
Molecular Weight (g/mol) 221.68
Synonym 6-Amino-6-(2-chlorophenyl)-2-cyclohexen-1-one, 5,6-Dehydronorketamine, Dehydronorketamine
IUPAC Name 6-amino-6-(2-chlorophenyl)cyclohex-2-en-1-one
SMILES NC1(CCC=CC1=O)c2ccccc2Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H12ClNO/c13-10-6-2-1-5-9(10)12(14)8-4-3-7-11(12)15/h1-3,5-7H,4,8,14H2
Formula Weight 221.0607
Chemical Name or Material Dehydronorketamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.