missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Tizanidine, TRC

Product Code. 30350999
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350999 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350999 Supplier Toronto Research Chemicals Supplier No. TRCD2305601MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 125292-34-4
Molecular Formula C9 H6 Cl N5 S
Quantity 1 mg
Synonym 2,1,3-Benzothiadiazol-4-amine, 5-chloro-N-1H-imidazol-2-yl-, 5-Chloro-N-1H-imidazol-2-yl-2,1,3-benzothiadiazol-4-amine, Dehydro tizanidine, Dehydrotizanidine
InChI Formula InChI=1S/C9H6ClN5S/c10-5-1-2-6-8(15-16-14-6)7(5)13-9-11-3-4-12-9/h1-4H,(H2,11,12,13)
SMILES Clc1ccc2nsnc2c1Nc3ncc[nH]3
IUPAC Name 5-chloro-N-(1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine
Molecular Weight (g/mol) 251.7
Formula Weight 251.0032
Chemical Name or Material dehydro Tizanidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.