Learn More
Delta2-Ceftazidime (>80%), TRC
Product Code. 30354223
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354223 2.5 mg 2.50mg
30350628 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30354223 Supplier Toronto Research Chemicals Supplier No. TRCC2441102.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1000980-60-8
Molecular Weight (g/mol) 546.58
Quantity 2.5 mg
Shipping Condition Room Temperature
Synonym Pyridinium, 1-[[(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-en-3-yl]methyl]-, inner salt∣Ceftazidime Delta-3-Isomer