missing translation for 'onlineSavingsMsg'
Learn More

Dembrexine EP Impurity C, Mikromol™

Product Code. 30051704
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051704 25 mg 25mg
30051031 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051704 Supplier Mikromol Supplier No. MM1200.030025
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dembrexine EP Impurity C
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dibromo-2-hydroxybenzaldehyde
CAS 90-59-5
Analyte or Component Names 3,5-Dibromo-2-hydroxybenzaldehyde
Shipping Condition Room Temperature
Molecular Formula C7 H4 Br2 O2
Certifications/Compliance ISO 17025
Synonym Dembrexine Hydrochloride Monohydrate Imp. C (EP), Dembrexine Imp. C (EP), 3,5-Dibromo-2-hydroxybenzaldehyde, Dembrexine Hydrochloride Monohydrate Impurity C, Dembrexine Impurity C
InChI Formula InChI=1S/C7H4Br2O2/c8-5-1-4(3-10)7(11)6(9)2-5/h1-3,11H
SMILES Oc1c(Br)cc(Br)cc1C=O
IUPAC Name 3,5-dibromo-2-hydroxybenzaldehyde
Molecular Weight (g/mol) 279.91
Quantity 25 mg
Formula Weight 277.8578 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.