missing translation for 'onlineSavingsMsg'
Learn More

Demeton (O&S), TRC

Product Code. 30134823
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134823 1 g 1g
30007612 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134823 Supplier Toronto Research Chemicals Supplier No. TRCD2314951G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8065-48-3
Molecular Formula 2 C8 H19 O3 P S2
Molecular Weight (g/mol) 516.68
Synonym Phosphorothioic acid, O,O-diethyl S-[2-(ethylthio)ethyl] ester, mixt. contg. (9CI), Bayer 8169, Demeton, E 1059, Ethyl systox, Mercaptofos, Mercaptophos, Phosphorothioic acid O,O-diethyl O-[2-(ethylthio)ethyl] ester mixture with O,O-diethyl S-[2-(ethylthio)ethyl]phosphorothioate, Septox, Systox
IUPAC Name diethoxy-(2-ethylsulfanylethoxy)-sulfanylidene-λ^{5}-phosphane;1-diethoxyphosphorylsulfanyl-2-ethylsulfanylethane
SMILES CCOP(=S)(OCC)OCCSCC.CCOP(=O)(OCC)SCCSCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C8H19O3PS2/c1-4-10-12(9,11-5-2)14-8-7-13-6-3;1-4-9-12(13,10-5-2)11-7-8-14-6-3/h2*4-8H2,1-3H3
Formula Weight 516.1026
Chemical Name or Material Demeton (O+S)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.