Learn More
Dermatan Sulfate (>90%), TRC
Product Code. 30010193
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010193 5 mg 5mg
30135096 25 mg 25mg
30007818 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010193 Supplier Toronto Research Chemicals Supplier No. TRCD2893255MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54328-33-5
Synonym Chondroitin Sulfate B Sodium Salt, Chondroitin Sulfate Type B Sodium Salt, Dermatan Sulfate Sodium Salt;
IUPAC Name sodium (3S,6R)-6-(((2S,5S)-3-acetamido-2-hydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)tetrahydro-2H-pyran-4-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C14H23NO15S.2Na/c1-3(17)15-5-10(9(30-31(24,25)26)4(2-16)27-13(5)23)28-14-8(20)6(18)7(19)11(29-14)12(21)22;;/h4-11,13-14,16,18-20,23H,2H2,1H3,(H,15,17)(H,21,22)(H,24,25,26);;/q;2*+1/p-2/t4?,5?,6?,7-,8?,9+,10?,11?,13-,14+;;/m0../s1
Chemical Name or Material Chondroitin sulfate B sodium salt