missing translation for 'onlineSavingsMsg'
Learn More

Des-4-fluorobenzyl Mosapride, TRC

Product Code. 30138567
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138567 1 mg 1mg
30132512 2.5 mg 2.5mg
30003753 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138567 Supplier Toronto Research Chemicals Supplier No. TRCD2899601MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152013-26-8
Molecular Formula C14 H20 Cl N3 O3
Molecular Weight (g/mol) 313.78
Synonym Benzamide, 4-amino-5-chloro-2-ethoxy-N-(2-morpholinylmethyl)-, Benzamide, 4-amino-5-chloro-2-ethoxy-N-(2-morpholinylmethyl)-, (±)-, 4-Amino-5-chloro-2-ethoxy-N-(2-morpholinylmethyl)benzamide, Des-4-fluorobenzyl mosapride, Des-p-fluorobenzyl mosapride
IUPAC Name 4-amino-5-chloro-2-ethoxy-N-(morpholin-2-ylmethyl)benzamide
SMILES CCOc1cc(N)c(Cl)cc1C(=O)NCC2CNCCO2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H20ClN3O3/c1-2-20-13-6-12(16)11(15)5-10(13)14(19)18-8-9-7-17-3-4-21-9/h5-6,9,17H,2-4,7-8,16H2,1H3,(H,18,19)
Formula Weight 313.1193
Chemical Name or Material des-4-fluorobenzyl Mosapride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.