missing translation for 'onlineSavingsMsg'
Learn More

Desacetyl Cefathiamidine, TRC

Product Code. 17903165
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17903165 50 mg 50mg
17901155 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17903165 Supplier Toronto Research Chemicals Supplier No. TRCD28827550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 958001-61-1
Molecular Formula C17 H26 N4 O5 S2
Molecular Weight (g/mol) 430.54
Synonym (6R,7R)-3-(Hydroxymethyl)-7-[[2-[[[(1-methylethyl)amino][(1-methylethyl)imino]methyl]thio]acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
IUPAC Name (6R,7R)-7-[[2-[(E)-N,N'-diisopropylcarbamimidoyl]sulfanylacetyl]amino]-3-(hydroxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(C)N\C(=N/C(C)C)\SCC(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H26N4O5S2/c1-8(2)18-17(19-9(3)4)28-7-11(23)20-12-14(24)21-13(16(25)26)10(5-22)6-27-15(12)21/h8-9,12,15,22H,5-7H2,1-4H3,(H,18,19)(H,20,23)(H,25,26)/t12-,15-/m1/s1
Formula Weight 430.1345
Chemical Name or Material Desacetyl Cefathiamidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.