missing translation for 'onlineSavingsMsg'
Learn More

Desacetyl Metipranolol Hydrochloride, TRC

Product Code. 30010710
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010710 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010710 Supplier Toronto Research Chemicals Supplier No. TRCD28810025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22664-59-1
Molecular Formula C15 H25 N O3 . Cl H
Quantity 25 mg
Synonym Phenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-2,3,6-trimethyl-, hydrochloride (9CI), 2-Propanol, 1-(4-hydroxy-2,3,5-trimethylphenoxy)-3-(isopropylamino)-, hydrochloride (8CI), VUFB 6502, 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-2,3,6-trimethylphenol hydrochloride, Deacetylmetipranolol hydrochloride, Deacetyltrimepranol hydrochloride, Desacetylmetipranolol hydrochloride, 1-(4-Hydroxy-2,3,5-trimethylphenoxy)-3-(isopropylamino)-2-propanol Hydrochloride
InChI Formula InChI=1S/C15H25NO3.ClH/c1-9(2)16-7-13(17)8-19-15-10(3)6-14(18)11(4)12(15)5;/h6,9,13,16-18H,7-8H2,1-5H3;1H
SMILES Cl.CC(C)NCC(O)COc1c(C)cc(O)c(C)c1C
IUPAC Name 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]-2,3,5-trimethylphenol;hydrochloride
Molecular Weight (g/mol) 303.8248
Formula Weight 303.1601
Chemical Name or Material Desacetyl Metipranolol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.