missing translation for 'onlineSavingsMsg'
Learn More

Desbenzyl Donepezil Hydrochloride, TRC

Product Code. 30008050
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008050 10 mg 10mg
30011860 100 mg 100mg
30009187 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008050 Supplier Toronto Research Chemicals Supplier No. TRCD28877510MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120013-39-0
Molecular Formula C17 H23 N O3 . Cl H
Molecular Weight (g/mol) 325.83
Synonym 2,3-Dihydro-5,6-dimethoxy-2-[(4-piperidinyl)methyl]indan-1-one Hydrochloride
IUPAC Name 5,6-dimethoxy-2-(piperidin-4-ylmethyl)-2,3-dihydroinden-1-one;hydrochloride
SMILES Cl.COc1cc2CC(CC3CCNCC3)C(=O)c2cc1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H23NO3.ClH/c1-20-15-9-12-8-13(7-11-3-5-18-6-4-11)17(19)14(12)10-16(15)21-2;/h9-11,13,18H,3-8H2,1-2H3;1H
Formula Weight 325.1445
Percent Purity >95%
Chemical Name or Material 2,3-Dihydro-5,6-dimethoxy-2-[(4-piperidinyl)methyl]indan-1-one Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.