missing translation for 'onlineSavingsMsg'
Learn More

Deschloro Florfenicol, TRC

Product Code. 30366355
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366355 25 mg 25mg
30359517 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30366355 Supplier Toronto Research Chemicals Supplier No. TRCD28879825MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138872-73-8
Molecular Formula C12 H15 Cl F N O4 S
Molecular Weight (g/mol) 323.77
Synonym Acetamide, 2-chloro-N-[(1S,2R)-1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]-, Acetamide, 2-chloro-N-[1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]-, [R-(R*,S*)]-, 2-Chloro-N-[(1S,2R)-1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]acetamide, Florfenicol-deschloro
IUPAC Name 2-chloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@@H](CF)NC(=O)CCl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H15ClFNO4S/c1-20(18,19)9-4-2-8(3-5-9)12(17)10(7-14)15-11(16)6-13/h2-5,10,12,17H,6-7H2,1H3,(H,15,16)/t10-,12-/m1/s1
Formula Weight 323.0394
Percent Purity >95%
Chemical Name or Material Deschloro Florfenicol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.