missing translation for 'onlineSavingsMsg'
Learn More

Desethylene Aripiprazole, TRC

Product Code. 30137507
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137507 25 mg 25mg
30136405 50 mg 50mg
30003387 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137507 Supplier Toronto Research Chemicals Supplier No. TRCD28912525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216394-63-6
Molecular Formula C21 H25 Cl2 N3 O2
Molecular Weight (g/mol) 422.35
Synonym Aripiprazole Desethylene Impurity
IUPAC Name 7-[4-[2-(2,3-dichloroanilino)ethylamino]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES Clc1cccc(NCCNCCCCOc2ccc3CCC(=O)Nc3c2)c1Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H25Cl2N3O2/c22-17-4-3-5-18(21(17)23)25-12-11-24-10-1-2-13-28-16-8-6-15-7-9-20(27)26-19(15)14-16/h3-6,8,14,24-25H,1-2,7,9-13H2,(H,26,27)
Formula Weight 421.1324
Chemical Name or Material Desethylene Aripiprazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.