missing translation for 'onlineSavingsMsg'
Learn More

Desfluoro Bicalutamide, TRC

Product Code. 30355686
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355686 1 mg 1mg
30349251 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30355686 Supplier Toronto Research Chemicals Supplier No. TRCD2899551MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90357-05-4
Molecular Formula C18H15F3N2O4S
Molecular Weight (g/mol) 412.38
Synonym Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methyl-3-(phenylsulfonyl)-, Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methyl-3-(phenylsulfonyl)-, (±)-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methyl-3-(phenylsulfonyl)propanamide, (2RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methyl-3-(phenylsulfonyl)propanamide, Desfluoro Bicalutamide
IUPAC Name 3-(benzenesulfonyl)-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide
SMILES CC(O)(CS(=O)(=O)c1ccccc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H15F3N2O4S/c1-17(25,11-28(26,27)14-5-3-2-4-6-14)16(24)23-13-8-7-12(10-22)15(9-13)18(19,20)21/h2-9,25H,11H2,1H3,(H,23,24)
Formula Weight 412.0705
Chemical Name or Material Desfluoro Bicalutamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.