missing translation for 'onlineSavingsMsg'
Learn More

Desfluoro Ezetimibe, TRC

Product Code. 30040751
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040751 25 mg 25mg
30040406 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30040751 Supplier Toronto Research Chemicals Supplier No. TRCD28994525MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302781-98-2
Molecular Formula C24 H22 F N O3
Molecular Weight (g/mol) 391.43
Synonym 2-Azetidinone, 3-[(3S)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-1-phenyl-, (3R,4S)-, Desfluoro Ezetimibe, Etezimibe Desfluoroaniline analog (USP)
IUPAC Name (3R,4S)-3-[(3S)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-1-phenylazetidin-2-one
SMILES O[C@@H](CC[C@@H]1[C@H](N(C1=O)c2ccccc2)c3ccc(O)cc3)c4ccc(F)cc4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H22FNO3/c25-18-10-6-16(7-11-18)22(28)15-14-21-23(17-8-12-20(27)13-9-17)26(24(21)29)19-4-2-1-3-5-19/h1-13,21-23,27-28H,14-15H2/t21-,22+,23-/m1/s1
Formula Weight 391.1584
Percent Purity >95%
Chemical Name or Material Desfluoro Ezetimibe
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.