missing translation for 'onlineSavingsMsg'
Learn More

Desfuroyl Ceftiofur Cysteine Disulfide (Contain 5% Ceftiofur) (>90%), TRC

Product Code. 17989724
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17989724 10 mg 10mg
17939534 1 mg 1mg
17937704 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17989724 Supplier Toronto Research Chemicals Supplier No. TRCD28990510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158039-15-7
Molecular Formula C17H20N6O7S4
Molecular Weight (g/mol) 548.64
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[[(2R)-2-amino-2-carboxyethyl]dithio]methyl]-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(2-amino-2-carboxyethyl)dithio]methyl]-7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-8-oxo-, [6R-[3(R*),6α,7β(Z)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[[(2R)-2-amino-2-carboxyethyl]dithio]methyl]-7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-8-oxo-, (6R,7R)- (9CI), (6R,7R)-3-[[[(2R)-2-Amino-2-carboxyethyl]dithio]methyl]-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, Desfuroylceftiofur cysteine disulfide, Desfuroyl ceftiofur cysteine disulfide
SMILES CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CSSC[C@H](N)C(=O)O)\c3csc(N)n3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H20N6O7S4/c1-30-22-9(8-5-32-17(19)20-8)12(24)21-10-13(25)23-11(16(28)29)6(2-31-14(10)23)3-33-34-4-7(18)15(26)27/h5,7,10,14H,2-4,18H2,1H3,(H2,19,20)(H,21,24)(H,26,27)(H,28,29)/b22-9+/t7-,10+,14+/m0/s1
Chemical Name or Material Desfuroyl Ceftiofur Cysteine Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.