missing translation for 'onlineSavingsMsg'
Learn More

Desisobutyl Febuxostat, TRC

Product Code. 30137878
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137878 100 mg 100mg
30133870 500 mg 500mg
30011706 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137878 Supplier Toronto Research Chemicals Supplier No. TRCD291100100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407582-48-3
Molecular Formula C12 H8 N2 O3 S
Molecular Weight (g/mol) 260.27
Synonym 2-(3-Cyano-4-hydroxyphenyl)-4-methyl-5-thiazolecarboxylic Acid, Desisobutyl Febuxostat
IUPAC Name 2-(3-cyano-4-hydroxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES Cc1nc(sc1C(=O)O)c2ccc(O)c(c2)C#N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H8N2O3S/c1-6-10(12(16)17)18-11(14-6)7-2-3-9(15)8(4-7)5-13/h2-4,15H,1H3,(H,16,17)
Formula Weight 260.0256
Chemical Name or Material Desisobutyl Febuxostat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.